Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class E2 with Certificate
Codice prodotto. 15595069 Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Peso (sistema metrico):
1 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Dimensione della confezione:
1 unità
Codice prodotto. 15595069
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
  • Class E2 stainless steel calibration weight
  • In compliance with OIML R111
  • Delivered with a calibration certificate guaranteeing traceability
Specifica
Classificazione E2
Materiale Stainless Steel
Peso (sistema metrico) 5 kg
Massa 5 kg
Forma Cylindrical