Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F2 with certificate
Codice prodotto. 15595179 Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Peso (sistema metrico):
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Dimensione della confezione:
1 unità
Codice prodotto. 15595179
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Use with weighing instruments of high accuracy class II. Fisherbrand™ Class F2 Stainless Steel Calibration Weights are also intended for use in important commercial transactions, such as those involving precious metals and stones.
  • Class F2 stainless steel calibration weight
  • Weights intended for use in the verification or calibration of class M1 and possibly class M2 weights
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
Specifica
Classificazione F2
Materiale Stainless Steel
Peso (sistema metrico) 5 kg
Massa 5 kg
Forma Cylindrical