Learn More
Invitrogen™ Influenza A NP Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA532242
Descrizione
Recommended positive controls: H1N1-infected H1299, DF1 (Influenza A virus treatment for 10hr), influenza virus (A/WSN/33) infected MDCK. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The nucleoprotein (NP) of Influenza A virus encapsidates the negative strand of the viral RNA and is essential for replicative transcription. It may also be involved in other essential functions throughout the virus life cycle. As well as binding ssRNA, NP is able to self associate to form large oligomeric complexes. NP is able to interact with a variety of other macromolecules of both viral and cellular origins. It binds the PB1 and PB2 subunits of the polymerase and the matrix protein M1. 'NP has also been shown to interact with at least four cellular polypeptide families: nuclear import receptors of the importin class, filamentous (F) actin, the nuclear export receptor CRM1 and a DEAD box helicase BAT1/UAP56' (Portela et al 2002).
Specifica
| Influenza A NP | |
| Polyclonal | |
| Unconjugated | |
| Influenza nucleoprotein | |
| Recombinant protein of Influenza A virus Nucleoprotein (A/WSN/1933(H1N1)). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.23 mg/mL | |
| PBS with 20% glycerol, 1% BSA and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Sicurezza e movimentazione
- Influenza A NP Polyclonal Antibody
Avvertenza
- Attenzione
Categoria di pericolo
- Lesione oculare grave/irritazione oculare Categoria 2
Indicazioni di pericolo
- H319-Provoca grave irritazione oculare.
Consigli di prudenza
- P260-Non respirare la polvere/i fumi/i gas/la nebbia/i vapori/gli aerosol.
- P264-Lavare accuratamente dopo l'uso.
- P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
Informazioni supplementari
- MIXTURE LIST-Contiene: glycerol
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.