Learn More
Thermo Scientific™ LinerGOLD™ GC Liners
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
Use the gold-standard Thermo Scientific™ LinerGOLD™ GC Liners for consistently high accuracy, sensitivity, and precision in your gas chromatography (GC) analysis.
Use the gold standard in GC liner performance and consistency. LinerGOLD GC Liners provide enhanced inertness, leading to increased accuracy, sensitivity, and precision in your gas chromatography (GC) analysis. These injection port liners deliver:
- Increased accuracy and sensitivity in analysis, preventing the loss of sensitive compounds in analysis
- Lower detection limits when analyzing active and sensitive compounds
- Excellent reproducibility gives confidence in your results from liner to liner
- Consistently high level of performance time after time
LinerGOLD GC liners are manufactured and tested to ensure that you have confidence with every liner that it installed onto your GC system.
Specifica
Specifica
| Quantità | 5 |
| Lunghezza (sistema metrico) | 78.5 mm |
| Materiale di impaccamento | None |
| Product Type | Double Taper Cyclo Liner |
| Diametro (sistema metrico) interno | 4 mm |
| Diametro (sistema metrico) esterno | 6.5 mm |
| Da utilizzare con (apparecchiatura) | TRACE 1300 Series and Agilent Instruments |