Learn More
Smeg™ Injection Trolley for Laboratory Glassware Washers
Codice prodotto. 11915594
Click to view available options
Capacità:
20 items
40 items
Include:
20 spigots, drying system connection
40 spigots, drying system connection
Da utilizzare con (applicazione):
Suitable for washing and drying narrownecked glassware up to 19.3 inches / 490 mm in height.
Washing narrow-necked glassware up to 490mmH
Dimensione della confezione:
Pezzo
Codice prodotto. 11915594
Marca: Smeg™ LM20DS
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Use these trolleys with Smeg™ glassware washers. Smeg Injection Trolleys for Laboratory Glassware Washers are available in a variety of configurations to fit a wide range of applications.
  • For use with Smeg glassware washers
  • Available with one, two and mixed spraying/injection levels
  • Clean and sanitize a wide variety of labware
Specifica
Materiale Stainless Steel
Product Type Injection Trolley
Capacità 20 items
Tipo di rack 20-position universal flask washing trolley in stainless steel with 20 spigots and drying system connection. Half the trolley space is free for mixed glassware suitable for a ½ basket accessory.
Stile Injection trolley with one spraying/injection level
Da utilizzare con (applicazione) Suitable for washing and drying narrownecked glassware up to 19.3 inches / 490 mm in height.
Da utilizzare con (apparecchiatura) Glassware washers
Include 20 spigots, drying system connection
Quantità 1 Each