missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha-5 Rabbit anti-Human, Mouse, Non-human primate, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10569-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Integrin alpha-5, also known as FNRA or VLA5A, is a protein that in humans is encoded by the ITGA5 gene. The product of this gene belongs to the integrin alpha chain family. Integrins are integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signaling.Specifica
| Integrin alpha-5 | |
| Polyclonal | |
| Unconjugated | |
| ITGA5 | |
| CD49e, FNRA, Integrin alpha 5, Integrin alpha F, ITGA5, VLA 5, VLA5A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16402, 3678 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P08648, P11688 | |
| ITGA5 | |
| Integrin alpha-5 Fusion Protein Ag0860 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Non-human Primate | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto