missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 2/CD18 Antibody (MEM-48), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB500-379APC
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Integrin beta 2/CD18 Monoclonal specifically detects Integrin beta 2/CD18 in Human samples. It is validated for Flow Cytometry.
Specifica
| Integrin beta 2/CD18 | |
| Monoclonal | |
| APC | |
| 959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody MEM-48 recognizes an epitope involving residues 534-546 in cysteine-rich repeat 3 of the CD18 antigen (integrin beta2 subunit; beta2 integrin). CD18 is a 90-95 kDa type I transmembrane protein expressed on all leukocytes. | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| MEM-48 | |
| Flow Cytometry | |
| ITGB2 | |
| Leukocytes of a patient suffering from a LGL-type leukemia. | |
| 0.1 mL | |
| Apoptosis, Cytokine Research, Immunology, Phospho Specific | |
| 3689 | |
| Human | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto