missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Intestinal Alkaline Phosphatase Polyclonal Antibody, HRP

Goat Polyclonal Antibody
Marca: Invitrogen™ PA126878
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PA1-26878 detects calf intestinal Alkaline Phosphatase from bovine samples.
There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The intestinal alkaline phosphatase gene encodes a digestive brush-border enzyme. This enzyme is upregulated during small intestinal epithelial cell differentiation.
Specifica
| Intestinal Alkaline Phosphatase | |
| Polyclonal | |
| HRP | |
| ALPI | |
| 2010001C14Rik; Akp2; Akp6; Alkaline phosphatase 1, intestinal, defined by SSR; alkaline phosphatase 6; alkaline phosphatase, intestinal; alkaline phosphomonoesterase; Alp1; ALPI; AP; EC 3.1.3.1; gIAP; glycerophosphatase; I Kasahara isozyme; IAP; IAP-1; IAP-I; intestinal alkaline phosphatase; intestinal alkaline phosphatase 1; intestinal alkaline phosphatase I IAP-I; intestinal alkaline phosphatase III; intestinal alkaline phosphatase IV; intestinal alkaline phosphatase V; intestinal-type alkaline phosphatase; intestinal-type alkaline phosphatase 1; Kasahara isozyme; PLAP-1; Regan isozyme; S51097 | |
| Goat | |
| IgG fraction | |
| RUO | |
| 280993 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Dot Blot, ELISA, Western Blot, Western Blot | |
| 10 mg/mL | |
| PBS with 1% BSA and 0.01% gentamicin sulfate; pH 7.2 | |
| P19111 | |
| ALPI | |
| Alkaline Phosphatase collected from calf intestine. | |
| 1 mg | |
| Primary | |
| Bovine | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto