Learn More
Brand™ Blaubrand™ IntraEND™ Disposable Micropipets
Sfoglia Tutto Brand Prodotti
Click to view available options
:
Capacity: 3μL
Descrizione
- Comprised of disposable glass without ring mark.
- Complies with DIN ISO 7550.
- CE Marked in accordance with IVD-Directive 98/79
- Calibrated to contain (TC, In.)
- Volume is defined by end-to-end filling
- Volumes greater than 5μL include DE-M marking.
Specifica
Specifica
| Accuracy | ≤±0.5% |
| Calibrazione | To Contain |
| Certificazione/Conformità | DIN ISO 7550, IVD-Directive 98/79 CE |
| Materiale | Glass |
| Quantità | 250/Box |
| Volume (sistema metrico) | 3 μL |
| Lunghezza (sistema metrico) | 32 mm |
| Monouso | Single-use |