Learn More
TERUMO Intravenous Catheters with 8SRD-codes
Click to view available options
Lunghezza (sistema metrico):
32mm
Dimensione della confezione:
pezzi 500
Descrizione
- A sterile single use, closure device
- Preventing clot formation during intermittent infusion or aspiration
- The large range in catheters and related obturators allows most appropriate choice of product for a wide range of applications
- Serves to close the catheter and to prevent clot formation during intermittent infusion
- An obturator with the same colour code as the catheter is available for each size
Specifica
Specifica
| Colore | Green |
| Senza lattice | Yes |
| Lunghezza (sistema metrico) | 32mm |
| Product Type | Obturator |
| Monouso | Single-use |
| Da utilizzare con (apparecchiatura) | IV Catheter |