missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRE1/ERN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27528-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IRE1 alpha is the endoplasmic reticulum to nucleus signalling 1 protein, a human homologue of the yeast Ire1 gene product. IRE1 alpha protein possesses intrinsic kinase activity and an endoribonuclease activity and it is important in altering gene expression as a response to endoplasmic reticulum-based stress signals. Specifically, IRE1 acts as the sensor of unfolded proteins in the ER that initiates transmittance of the unfolded protein signal from the ER to the nucleus by splicing XBP1 mRNA converting it into a potent unfolded-protein response transcriptional activator. Null mutants are viable, but IRE1 is essential for viability under stress conditions that cause unfolded proteins to accumulate in the ER. Depletion of IRE1p through the expression of a domit negative form of IRE1p has no effect on transfected cells, but cell death via apoptosis occurs under stress conditions that cause unfolded proteins to accumulate in the ER.Specifica
| IRE1/ERN1 | |
| Polyclonal | |
| Unconjugated | |
| ERN1 | |
| ERN1, hIRE1p, Inositol requiring protein 1, IRE1, Ire1 alpha, IRE1P | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2081 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75460 | |
| ERN1 | |
| IRE1; ERN1 Fusion Protein Ag25580 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto