missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11312-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
TYRO3 (tyrosine-protein kinase receptor TYRO3) is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding to several ligands including TULP1 or GAS6. It regulates many physiological processes including cell survival, migration, and differentiation. Ligand binding at the cell surface induces dimerization and autophosphorylation of TYRO3 on its intracellular domain that provides docking sites for downstream signaling molecules. Following activation by a ligand, TYRO3 plays a role in various processes such as neuron protection from excitotoxic injury, platelet aggregation, and cytoskeleton reorganization.Specifica
| IRF3 | |
| Polyclonal | |
| Unconjugated | |
| IRF3 | |
| interferon regulatory factor 3, IRF 3, IRF3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 292892, 3661, 54131 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Flow Cytometry, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P70671, Q14653 | |
| Irf3 | |
| IRF3 Fusion Protein Ag1858 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto