Learn More
Iron(II) fumarate, 94%
Codice prodotto. p-7027303
Click to view available options
Quantità:
250 g
1000 g
Dimensione della confezione:
1000g
250g
Codice prodotto. 11436836
Marca: Thermo Scientific Alfa Aesar B23956.30
Richiesta di formato massivo o personalizzato
CAS: 141-01-5 | C4H2FeO4-2 | 169.901 g/mol
Iron(II) fumarate is used as dietary Supplement, nutrient, iron source in foods and pharmaceuticals, as animal feed additive and in infant formulas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Iron(II) fumarate is used as dietary Supplement, nutrient, iron source in foods and pharmaceuticals, as animal feed additive and in infant formulas.
Solubility
Soluble in water (0.14 g/100 ml at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 141-01-5
Formula molecolare C4H2FeO4-2
Molecular Weight (g/mol) 169.901
Numero MDL MFCD00058315
InChI Key OOPLWEDSEDELIX-TYYBGVCCSA-L
Sinonimo ferrous fumarate, eisenfumarat
PubChem CID 66524104
IUPAC Name (E)-but-2-enedioate;iron
SMILES C(=CC(=O)[O-])C(=O)[O-].[Fe]
Specifica
Materiale o nome chimico Iron(II) fumarate
Punto di fusione >300°C
Quantità 250 g
Indice di Merck 14,4046
Informazioni di solubilità Soluble in water (0.14g/100 ml at 25°C).
Peso formulazione 169.9
Percent Purity 94%
Densità 2.44