missing translation for 'onlineSavingsMsg'
Learn More

Itaconic acid, 99%

Codice prodotto. p-7022679
Click to view available options
Quantità:
250 g
1000 g
5000 g
Dimensione della confezione:
1000g
250g
5000g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 11453027

Marca: Thermo Scientific Alfa Aesar A15566.30

Richiesta di formato massivo o personalizzato

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 97-65-4 | C5H6O4 | 130.099 g/mol

Itaconic acid is used in the preparation of acrylonitrile-butadiene-styrene and acrylate latexes. It is also used to prepare poly-itaconic acid, resins biofuel components and ionomer cements. It finds application in the textile, chemical and pharmaceutical industries. It is also used as an additive in fibers and ion exchange resins to increase abrasion, waterproofing, physical resistance, dying affinity and better duration. Further, it acts as a co-monomer used in the preparation of acrylic fibers and rubbers, reinforced glass fiber, artificial diamonds and lens. In addition to this, it acts as a binder and sizing agent in non-weaving fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Itaconic acid is used in the preparation of acrylonitrile-butadiene-styrene and acrylate latexes. It is also used to prepare poly-itaconic acid, resins biofuel components and ionomer cements. It finds application in the textile, chemical and pharmaceutical industries. It is also used as an additive in fibers and ion exchange resins to increase abrasion, waterproofing, physical resistance, dying affinity and better duration. Further, it acts as a co-monomer used in the preparation of acrylic fibers and rubbers, reinforced glass fiber, artificial diamonds and lens. In addition to this, it acts as a binder and sizing agent in non-weaving fibers.

Solubility
Soluble in water, acetone, methanol, hexane and ethanol. Slightly soluble in benzene, chloroform, carbon disulfide and petroleum ether.

Notes
Hygroscopic. Moisture sensitive. Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 97-65-4
Formula molecolare C5H6O4
Molecular Weight (g/mol) 130.099
Numero MDL MFCD00004260
InChI Key LVHBHZANLOWSRM-UHFFFAOYSA-N
Sinonimo itaconic acid, 2-methylenesuccinic acid, methylenesuccinic acid, methylenebutanedioic acid, propylenedicarboxylic acid, itaconate, 2-propene-1,2-dicarboxylic acid, butanedioic acid, methylene, succinic acid, methylene, 2-methylenebutanedioic acid
PubChem CID 811
ChEBI CHEBI:30838
IUPAC Name 2-methylidenebutanedioic acid
SMILES C=C(CC(=O)O)C(=O)O

Specifica

Materiale o nome chimico Itaconic acid
Punto di fusione 165°C to 169°C
Densità 1.63
Punti di ebollizione 268°C
Quantità 250 g
Beilstein 1759501
Sensibilità Hygroscopic
Indice di Merck 14,5242
Informazioni di solubilità Soluble in water,acetone,methanol,hexane and ethanol. Slightly soluble in benzene,chloroform,carbon disulfide and petroleum ether.
Peso formulazione 130.1
Percent Purity 99%
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Itaconic acid
Avvertenza
  • Pericolo
Categoria di pericolo
  • Lesione oculare grave/irritazione oculare Categoria 1
Indicazioni di pericolo
  • H318-Provoca gravi lesioni oculari.
Consigli di prudenza
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
  • P310-Contattare immediatamente un CENTRO ANTIVELENI/un medico .
Informazioni supplementari
  • MIXTURE LIST-Contiene: Itaconic acid

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato