Learn More
Invitrogen™ ITGB6 Monoclonal Antibody (437211), APC
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA523675
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ITGB6 Monoclonal Antibody for Flow
ITGB6 (Integrin alpha-V/beta-6) is a receptor for fibronectin and cytotactin. It recognizes the sequence R-G-D in its ligands. Internalization of integrin alpha-V/beta-6 via clathrin-mediated endocytosis promotes carcinoma cell invasion.
Specifica
| ITGB6 | |
| Monoclonal | |
| APC | |
| Itgb6 | |
| 2210409C20Rik; 4831415H04Rik; AI1H; integrin beta 6; integrin beta-6; integrin subunit beta 6; integrin, beta 6; Itgb6 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3694 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 437211 | |
| Buffered saline solution with ≤ 0.5% BSA and 0.09% sodium azide | |
| P18564 | |
| Itgb6 | |
| CHO-derived recombinant human Integrin alpha V beta 6 Phe31-Val992 (Integrin alpha V) and Gly22-Asn707 (Integrin beta 6). | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |