Learn More
GPE Scientific™ J Young Spherical Joint, Male/O-Ring Ball Joint
Click to view available options
Tipo:
Male Spherical Joint
Joint Size:
12/2
12/5
18/7
18/9
28/12
28/15
35/20
40/25
51/35
Dimensione della confezione:
Pezzo
Descrizione
- Spherical joint designed for use with cup joint
- Available with different joint sizes, shank bores, and shank ODs
Specifica
Specifica
| Descrizione articolo | Shank Bore: 2 mm, Shank OD: 7 mm |
| Tipo | Male Spherical Joint |
| Joint Size | 12/2 |