Learn More
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and O-Ring Ball Joint Cup
Codice prodotto. p-7147048
Click to view available options
Capacità (sistema metrico):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Dimensione della confezione:
Pezzo
Codice prodotto. 15894734
Marca: GPE Scientific™ CSR/25
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and O-Ring Ball Joint Cup is designed for use with Schlenk lines and a vacuum manifold and available in various capacities.
  • Designed with a side arm for evacuating or filling with gases
  • For connection to Schlenk lines
  • Round-bottomed shape
  • Includes O-ring tap
  • Includes O-ring ball joint cup
Specifica
Capacità (sistema metrico) 25 mL
Include O-Ring Tap and O-Ring Ball Joint Cup
Product Type Vacuum/Schlenk Flask
Giunto del braccio laterale Size: 19