missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JunD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA538138
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The activator protein-1 (AP-1) transcription factor consists of either Jun/Jun homodimers or Fos/Jun heterodimeric complexes. Homo- and heterodimers bind to the TGACTCA consensus sequence present in numerous promoters and initially identified as the phorbol ester tumor promoter response element (TRE). Jun B and Jun D have been shown to be almost identical to c-Jun in their C-terminal regions, which are involved in dimerization and DNA binding, whereas their N-terminal domains, which are involved in transcriptional activation, diverge. All three form heterodimers among themselves and with c-Fos and other members of the Fos gene family. Studies suggest that the two forms of Jun D may be due to internal initiation of translation.
Specifica
| JunD | |
| Polyclonal | |
| Unconjugated | |
| JUND | |
| Transcription factor jun-D, JUN D | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16478, 24518, 3727 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P17535, P15066, P52909 | |
| JUND | |
| A synthetic non-phosphopeptide derived from human JunD around the phosphorylation site of Ser255 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto