Learn More
Kartell™ K120 Rubber Hoses
Click to view available options
Diametro (sistema metrico) interno:
5 mm
6 mm
8 mm
Dimensione della confezione:
6m
Descrizione
- Includes cloth impression surface
- Suitable for air, water, medium concentration acids and alcaline solutions, abrasive products, oxygenized solvents etc.
Specifica
Specifica
| Diametro (sistema metrico) interno | 8 mm |
| Materiale | Gomma |
| Diametro (sistema metrico) esterno | 16 mm |
| Overall Length (Metric) | 6 m |
| Da utilizzare con (applicazione) | Suitable for use with air, water, medium concentration acids, alkaline solutions, abrasive products and oxygenized solvents |
| Min. Operating Temperature (Metric) | ° |
| Quantità confezione | 6 m |
| Max. Operating Temperature (Metric) | 70°C |