missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA578544
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Positive Control: SK-N-SH, mouse hippocampus, rat hippocampus, rat spinal cord Predicted Reactivity: Mouse (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Mediates electroneutral potassium-chloride cotransport in mature neurons. Transport occurs under isotonic conditions, but is activated 20-fold by cell swelling. Important for Cl(-) homeostasis in neurons.
Specifica
| KCC2 | |
| Polyclonal | |
| Unconjugated | |
| SLC12A5 | |
| EIEE34; EIG14; Electroneutral potassium-chloride cotransporter 2; erythroid K-Cl cotransporter 2; furosemide-sensitive K-Cl cotransporter; hKCC2; KCC 2; Kcc2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; K-Cl cotransporter 2; KIAA1176; mKCC2; mKIAA1176; neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; S12A5; SLC12A5; solute carrier family 12 (potassium/chloride transporter), member 5; solute carrier family 12 (potassium-chloride transporter), member 5; solute carrier family 12 member 5; solute carrier family 12, (potassium-chloride transporter) member 5; solute carrier family 12, member 5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 171373, 57138, 57468 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.36 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q63633, Q91V14, Q9H2X9 | |
| SLC12A5 | |
| Synthetic peptide encompassing a sequence within the Intracellular domain of mouse KCC2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto