missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA590504
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: TKNEREREIQ SITDESRGSI RRKNPANTRL RLNVPEETAG DSEEKPEEEV QLIHDQSAPS CPSSSPSPGE EPEGEGETDP E; Positive Samples: Mouse brain; Cellular Location: Membrane, Multi-pass membrane protein.
Mediates electroneutral potassium-chloride cotransport in mature neurons. Transport occurs under isotonic conditions, but is activated 20-fold by cell swelling. Important for Cl(-) homeostasis in neurons.
Specifica
| KCC2 | |
| Polyclonal | |
| Unconjugated | |
| SLC12A5 | |
| EIEE34; EIG14; Electroneutral potassium-chloride cotransporter 2; erythroid K-Cl cotransporter 2; furosemide-sensitive K-Cl cotransporter; hKCC2; KCC 2; Kcc2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; K-Cl cotransporter 2; KIAA1176; mKCC2; mKIAA1176; neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; S12A5; SLC12A5; solute carrier family 12 (potassium/chloride transporter), member 5; solute carrier family 12 (potassium-chloride transporter), member 5; solute carrier family 12 member 5; solute carrier family 12, (potassium-chloride transporter) member 5; solute carrier family 12, member 5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57138 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q91V14 | |
| SLC12A5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 945-1025 of human SLC12A5 (NP_0011282431). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto