missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KChIP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14212-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This gene encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins (KCNIPs), which belong to the recoverin branch of the EF-hand superfamily. Members of the KCNIP family are small calcium binding proteins. They all have EF-hand-like domains, and differ from each other in the N-terminus. They are integral subunit components of native Kv4 channel complexes. They may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. Alternative splicing results in multiple transcript variant encoding different isoforms.Specifica
| KChIP1 | |
| Polyclonal | |
| Unconjugated | |
| KCNIP1 | |
| KCHIP1, KCNIP1, VABP, Vesicle APC binding protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 30820 | |
| -20°C | |
| Liquid |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NZI2 | |
| KCNIP1 | |
| KChIP1 Fusion Protein Ag5432 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto