Learn More
DWK Life Sciences™ KECK™ Adapter KA, complete, from hose connection glass to connection plastic
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Size:
GL 14, diameter 4.5 mm
GL 14, diameter 9.0 mm
RD 14, diameter 4.5 mm
RD 14, diameter 9.0 mm
Dimensione della confezione:
pezzi 10
Descrizione
KECK™ adapters prevent glass breakage on olives and protect fingers and hands from injuries. The clamping ring, O-ring and screw cap are permanently screwed to the glass olive. The hose part can be connected quickly, securely and tightly to the threaded cap with just 1/4 turn and can also be loosened again just as quickly.
- Quick connection and disconnection of the hose part to the threaded cap with only 1/4 turn
- permanent screw connection with the glass nozzle
- KECK™ adapters prevent glass breakage on nozzles and protect fingers and hands from injuries
Specifica
Specifica
| Quantità | 10 |
| Da utilizzare con (apparecchiatura) | Connection of a Hose Piece with a Threaded Cap |
| Da utilizzare con (applicazione) | Connection of a hose piece with a threaded cap |
| Size | GL 14, diameter 4.5 mm |