Learn More
DWK Life Sciences™ KECK™ Clip Assortment, for conical joints, from metal
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Da utilizzare con (apparecchiatura):
Secure Glass-to-Glass Joints
Dimensione della confezione:
Pezzo
Descrizione
KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).
- For conical joints
- Dimensionally stable with particularly good spring properties
- Easy, safe and quick to set up
Specifica
Specifica
| Quantità | 1 Ea. |
| Materiale | Metal |
| Include | 2 x KCM 14, 2 x KCM 19, 1 x KCM 29, 1 x KCM 24 |
| Da utilizzare con (apparecchiatura) | Secure Glass-to-Glass Joints |
| Da utilizzare con (applicazione) | Secure glass-to-glass joints |