Learn More
DWK Life Sciences™ KECK™ Clip for Conical Joints, Ni, KCM
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Size:
NS 14
NS 19
NS 24
NS 29
Dimensione della confezione:
Pezzo
pezzi 100
Descrizione
For applications at very high temperatures, the KECK™ nickel-plated spring steel ground joint clamps an important addition to KECK™ conical ground joint clips made of POM (polyoxymethylene). Compared to conventional wire clamps, they are easy and safe to attach and are inherently stable.
- Easy and safe to put on and are dimensionally stable
- Good dimensional stability compared to conventional wire clamps
Specifica
Specifica
| Quantità | 100 |
| Materiale | Nickel |
| Da utilizzare con (apparecchiatura) | For Use at Very High Temperatures |
| Da utilizzare con (applicazione) | For use at very high temperatures; Securing conical ground joints |
| Size | NS 24 |