Learn More
KG-501, MedChemExpress
Product Code. 16483818
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483818 10 mg 10mg
16466547 5 mg 5mg
16460197 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16483818 Supplier Medchem Express Supplier No. HY103299
 Restricted Item
Spedizione stimata: 23-11-2026
MedChemExpress KG-501 is a CREB inhibitor, with an IC50 of 6.89 μM.
KG-501 is a CREB inhibitor, with an IC50 of 6.89 μM.
Specifications
Chemical Name or Material KG-501
CAS 18228-17-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C17H13ClNO5P
Quantity 10 mg
Synonym Naphthol AS-E phosphate
Solubility Information DMSO : 6 mg/mL (15.88 mM; Need ultrasonic and warming)
SMILES O=C(C1=C(OP(O)(O)=O)C=C2C=CC=CC2=C1)NC3=CC=C(Cl)C=C3
Molecular Weight (g/mol) 377.72
Formula Weight 377.72
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only