missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ki67/MKI67 Antibody (8D5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 11 publications
292.00€
Specifica
| Antigene | Ki67/MKI67 |
|---|---|
| Clone | 8D5 |
| Concentrazione | 1.0 mg/mL |
| Diluizione | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100. Use reported by customer review, Immunohistochemistry-Paraffin 1:200-1:1000, Immunohistochemistry-Frozen reported in scientific literature (PMID 33131911), Knockout Validated |
| Applicazioni | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
18743184
|
Novus Biologicals
NBP2-22112SS |
0.025 mL |
292.00€
0.03mL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
Ki67/MKI67 Monoclonal specifically detects Ki67/MKI67 in Human, Mouse, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.Specifica
| Ki67/MKI67 | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
| P46013 | |
| 4288 | |
| The immunogen for this KI67/MKI67 Antibody (8D5) was made using a synthetic peptide from the internal region of Human KI67/MKI67 (amino acids: 1100-1113, 1222-1235 and 3 more spots); sequence: CEDLAGFKELFQTPG [Uniprot: P46013]. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 8D5 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100. Use reported by customer review, Immunohistochemistry-Paraffin 1:200-1:1000, Immunohistochemistry-Frozen reported in scientific literature (PMID 33131911), Knockout Validated | |
| Monoclonal | |
| Precipitation | |
| RUO | |
| Human, Rabbit | |
| antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
| MKI67 | |
| IgG1 | |
| Ammonium sulfate precipitation | |
| 359 kDa |
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto