Learn More
DWK Life Sciences Kimble™ KIMAX™ Class A Cassia Flask
Codice prodotto. 11813173
Click to view available options
:
Std. Taper Stopper, No.13; 100 to 110mL
Codice prodotto. 11813173
Marca: DWK Life Sciences 28066110
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Calibrated To Contain
Calibrated To Contain
  • Official U.S. Pharmacopeia method to assay cinnamaldehyde and to analyze cassia, cinnamon, and clove extracts
  • Determine amount of unsaturated constituents in straw oil or similar wash oils, by sulfonation method
  • Tolerance: ±0.08mL
  • Capacity: 100 to 110mL in 0.1mL divisions
  • With standard taper 13 stopper
TRUSTED_SUSTAINABILITY
Specifica
Calibrazione Calibrata per riempimento
Capacità (sistema metrico) 110 mL
Materiale Borosilicate Glass
Altezza (sistema imperiale) 10.23 in.
Altezza (sistema metrico) 26 cm
Tolleranza ±0.08 mL
Graduated Yes
Dimensione chiusura 13