Learn More
DWK Life Sciences Kimble™ KIMAX™ Pycnometer Bottle
Codice prodotto. p-4842611
Click to view available options
Capacità (sistema metrico):
25 mL
50 mL
Altezza (sistema metrico):
175 mm
186 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 12737567
Marca: DWK Life Sciences 15123N50
Designed for measuring and comparing the densities or specific gravities of liquids and solids. This non-mercury spirit-filled pycnometer is supplied with Standard Taper 10/18 thermometer with a range of 14 to 38°C in 0.2° increments.
  • Thermometer is retested for accuracy within ±0.2 degrees C
  • Standard Taper 5/12 side tube cap has a small vent near the top
  • Body is conical for maximum stability
  • Capacity is within 10 percent of nominal shown on the flask
  • Each flask body is numbered for ready identification
TRUSTED_SUSTAINABILITY
Specifica
Capacità (sistema metrico) 50 mL
Altezza (sistema metrico) 186 mm
Product Type Pycnometer Bottle