Learn More
DWK Life Sciences Kimble™ KIMAX™ Serialized/Certified Cylindrical Microflasks
Click to view available options
Capacità (sistema metrico):
1 mL
2 mL
5 mL
10 mL
25 mL
Dimensione della confezione:
pezzi 6
Descrizione
Calibrated To Contain
- Permanently marked with individual serial number
- Individually calibrated and certified against equipment whose calibration is traceable to NIST™
- Cylindrical body for better mixing and draining
- Easier to withdraw samples using pipets
- Wide base provides stability
- Marking spot
- Ground glass stopper
- 1 and 2mL sizes come with standard taper 8 stopper; 10mL size with standard taper 9
Compliance
Designed from requirements of ASTM™ E 237, Class A serialized.
Specifica
Specifica
| Calibrazione | Calibrata per riempimento |
| Materiale chiusura | Vetro |
| Capacità (sistema metrico) | 5 mL |
| Materiale | Borosilicate Glass |
| Altezza (sistema imperiale) | 3.9 in. |
| Altezza (sistema metrico) | 9.9 cm |
| Tolleranza | ±0.02 mL |
| Graduated | Yes |
| Dimensione chiusura | 8 |