Learn More
DWK Life Sciences Kimble™ Kontes™ Filtration Assemblies: Flasks With Side Arm, No. 2 Hose connection
Click to view available options
Capacità (sistema metrico):
125 mL
1000 mL
4000 mL
Giunto rastremato standard:
5
8
Dimensione della confezione:
Pezzo
Descrizione
DWK Life Sciences Kimble Kontes Filtration Assemblies: Flasks With Side Arm, No. 2 Hose connection, Connector Types: No. 2 Hose Connection, Borosilicate Glass, Description: Filtration Flask
Order Info
Does not include rubber stopper and hose
Specifica
Specifica
| Capacità (sistema metrico) | 4000 mL |
| Materiale | Vetro borosilicato |
| Include | No. 2 Hose Connection |
| Product Type | Filtration Flask |
| Giunto rastremato standard | 8 |
| Quantità | 1/Cs. |