Learn More
Thermo Scientific™ Kinase buffer I (5X)
Codice prodotto. 15434449
Cambia vista
Click to view available options
Quantità:
100 mL
250 mL
Dimensione della confezione:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
15434449 100 mL 100mL
15444449 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 15434449 Fornitore Thermo Scientific™ N. del fornitore J61967.AE
Richiesta di formato massivo o personalizzato
For cellular and molecular biology applications
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Formulation:100mM Tris-HCl 100, 50mM magnesium chloride, 5mM dithiothreitol, and 5mM sodium orthovanadate, pH 7.4
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Kinase buffer I (5X)
Forma fisica Liquid
Quantità 100 mL
Formulazione 100mM Tris-HCl 100, 50mM magnesium chloride, 5mM dithiothreitol, and 5mM sodium orthovanadate, pH 7.4.
For Research Use Only