Learn More
MEDICOM Kolmi Coverfeel Touch Blue Nitrile Glove
GREENER_CHOICE
Codice prodotto. p-200104845 Sfoglia Tutto Medicom Prodotti
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Dimensione della confezione:
pezzi 100
pezzi 90
Codice prodotto. 30215954
Marca: MEDICOM 1140A
Solo null disponibili
Aggiungi al carrello
Edge
Aggiungi al carrello
Great elasticity and optimal comfort. Excellent grip of objects. Recommended in medical, industrial, and agri-food environments. Made in France.
  • No accelerator to limit type IV allergies.
  • Ultra comfortable glove with faster donning thanks to its internal polymer coating.
  • Great elasticity and good dexterity.
  • Non-slip texture for excellent grip and object manipulation.
  • Recommended for medical, industrial, and agri-food environments.
Specifica
Chlorinated No
Accelerator Free Yes
Applicazioni Industrial, Healthcare, Laboratory
AQL 1.5
Colore Blue
Monouso Single-use
Finitura esterna Fingertip Textured
Finger Thickness (English) 3.14 mil
Finger Thickness (Metric) 0.08 mm
Latex Free
Materiale Nitrile
Palm Thickness (English) 2.7 mil
Palm Thickness (Metric) 0.07 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Small
Sterilità Non-sterile
Spessore (sistema imperiale) 2.7 mil
Spessore (sistema metrico) 0.07 mm
FDA 510 (k) No
Stile polso guanto Rolled
Lunghezza (sistema metrico) 240 mm