Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm)
Click to view available options
Volume (sistema metrico):
1000 mL (Funnel), 4000 mL (Flask)
300 mL (Funnel), 1000 mL (Flask)
500 mL (Funnel), 2000 mL (Flask)
Dimensione della confezione:
Pezzo
Descrizione
Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents
- The ground joint connection eliminates the phthalate contamination that can occur when using silicone or neoprene stoppers
- The support base has a 40-60 micron porosity glass frit and an integral vacuum connection that is located above the drip tip to prevent contamination of the vacuum line with filtrate droplets
- Each apparatus is supplied with a funnel, an anodized aluminum clamp, a 47 mm fritted glass support base and a filtration flask
- Joints are 40/35
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifica
Specifica
| Materiale | Glass |
| Tipo | Microfiltration Assembly |
| Volume (sistema metrico) | 1000 mL (Funnel), 4000 mL (Flask) |