Learn More
OHAUS™ Lab Lift
Sfoglia Tutto Ohaus Prodotti
Click to view available options
Size:
102 x 102 (L x W) Platform
152 x 152 (L x W) Platform
203 x 203 (L x W) Platform
254 x 254 (L x W) Platform
305 x 305 (L x W) Platform
406 x 406 (L x W) Platform
76 x 76 (L x W) Platform
Materiale:
Aluminum
Stainless Steel
Dimensione della confezione:
Pezzo
Descrizione
- Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
- Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
- Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
Specifica
Specifica
| Tipo | Lab Lift |
| Materiale | Stainless Steel |
| Descrizione | Clamp, Support, Lab-Lift, CLR-LBLFTS010 |
| Altezza (sistema metrico) | 64 to 127 mm |
| Size | 102 x 102 (L x W) Platform |