Learn More
Heat Shock Protein 70 (HSP70) Ab-2 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments.
Marca: Epredia MS-482-B1
Descrizione
Heat shock proteins (HSP's) or stress response proteins (SRP's) are synthesized in variety of environmental and pathophysiological stressful conditions. Many HSP's are involved in processes such as protein denaturation-renaturation, folding-unfolding, transport-translocation, activation-inactivation, and secretion. HSP70 is found to be associated with steroid receptors, actin, p53, polyoma T antigen, nucleotides, and other unknown proteins. Also, HSP70 has been shown to be involved in protective roles against thermal stress, cytotoxic drugs, and other damaging conditions.Host Species: Mouse
Clone: W27
Isotype: IgG2a
Species Reactivity: Human, Monkey and Cow. Others not known.
Epitope: Not determined
Immunogen: HSP70 protein from HeLa cells
Molecular Weight: 73 kDa (constitutive), 72 kDa (inducible)
Positive Control: BT474 or HeLa cells, Breast CA
Cellular Localization: Nuclear and cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifica
| Heat Shock Protein 70 (HSP70) Ab-2 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 500 μL | |
| Stress Response | |
| Bovine, Human, Non-human Primate | |
| IgG2a |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| W27 | |
| Biotin | |
| HSP70 protein from HeLa cells | |
| RUO | |
| Primary | |
| Concentrated |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.