Learn More
Kappa Light Chain Ab-1 Mouse Monoclonal Antibody, Epredia™
Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Kappa Light Chain Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Marca: Epredia MS-143-B1
Descrizione
Antibody to the κ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.Host Species: Mouse
Clone: L1C1
Isotype: IgG1/k
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Immunogen: B-lymphoma cells
Molecular Weight: 25kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifica
| Kappa Light Chain Ab-1 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 500 μL | |
| Hematopoietic | |
| Human | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| L1C1 | |
| Biotin | |
| B lymphoma cells | |
| IVD | |
| Primary | |
| Concentrated |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.