Learn More
Prohibitin (Mitochondrial Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Prohibitin (Mitochondrial Marker) Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia MS-261-PABX
Descrizione
Prohibitin is an evolutionarily conserved protein located in the inner membrane of mitochondria. Prohibitin appears to be a very reliable marker of mitochondria. Prohibitin shows antiproliferative activity and has been proposed to play a role in normal cell cycle regulation, replicative senescence, cellular immortalization, and tumor suppression.Host Species: Mouse
Clone: II-14-10
Isotype: IgG1
Species Reactivity: Human, Pig, Rat, Mouse, and Chicken. Others not known.
Epitope: Not determined
Immunogen: Purified recombinant rat prohibitin protein
Molecular Weight: 30kDa
Positive Control: MAD109 or PC12 cells, Heart
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifica
| Prohibitin (Mitochondrial Marker) Ab-1 | |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 200 μL | |
| Apoptosis | |
| Chicken, Human, Mouse, Pig, Rat | |
| IgG1 |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| II-14-10 | |
| Unconjugated | |
| Purified recombinant rat prohibitin protein | |
| RUO | |
| Primary | |
| Concentrated |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.