Learn More
Labconco™ CentriVap™ Solvent Systems
Sfoglia Tutto Labconco Corporation Prodotti
Click to view available options
Requisiti elettrici:
230 V 50 Hz
Tipo di spina:
International - Schuko
Certifications:
CE
Dimensione della confezione:
Pezzo
Descrizione
Includes
Includes: Benchtop concentrator with glass lid, cold trap, solvent insert, clear canister, 12-13mm rotor, vacuum tubing and six clamps
Combines multiple concentration components into one easy for ordering and just by adding vacuum pump it will be ready to concentrate samples.
- Benchtop style
- Lid type: glass
- Type/Application: solvents
- Accessories (Required): Diaphragm or Rotary Vane vacuum pump
Specifica
Specifica
| Modello | Centrivap |
| Certifications | CE |
| Frequenza | 50 Hz |
| Include | Benchtop concentrator with Glass lid, cold trap, solvent insert, clear canister, 12 to 13mm rotor, vacuum tubing and 6 clamp |
| Tipo | CentriVap System |
| Tensione | 230 V |
| Requisiti elettrici | 230 V 50 Hz |
| Amperaggio | 4 A |
| Tipo di spina | International - Schuko |