Descrizione
Freeze drying ampule accessories allow ampules to be used with Labconco FreeZone Freeze Dryers.
- Large rubber split stoppers fit 5, 10, 20, 30, 50, 100 and 125 ml Labconco Serum Bottles (bottles with 20 mm corkage).
- Package of 100.
- Oxygen/Natural Gas Sealing Torch is specifically designed for flame sealing freeze dry ampules.
- Seals all types of heat-resistant glass.
- Connects to natural gas, butane or propane and oxygen with 1/4 in. ID hose connectors.
- Ampule valve adapter provides connection of ampules to drying chambers and manifolds by connecting ampules to valve ports or stainless steel stems.
- Ten per package.
Specifica
Specifica
| Tipo | Stopper |
| Materiale | Rubber |
| Quantità | 100 Pk. |
| Da utilizzare con (apparecchiatura) | Freeze drying system |
| Diametro (sistema metrico) | 2 cm |