Learn More
Labconco™ SteamScrubber™ Labware Inserts
Hold a variety of wider-mouth flasks, beakers and other labware in inverted position
Descrizione
Hold a variety of wider-mouth flasks, beakers and other labware in inverted position
- Pins have plastic tips to protect glassware from scratching
- Retainer Top is recommended for use with plastic and lightweight glassware
- Type 304 stainless steel
- Fits standard bottom rack on SteamScrubber units only
Specifica
| Each |