Learn More
Buerkle™ LaboPlast™ Disposable Scoop
BIOPHARMAGREENER_CHOICE
Codice prodotto. p-7084546 Sfoglia Tutto Bürkle Prodotti
Click to view available options
Capacità (sistema metrico):
25 mL
50 mL
100 mL
150 mL
250 mL
500 mL
1000 mL
2000 mL
Include:
With Cover
Without Cover
Dimensione della confezione:
pezzi 10
Codice prodotto. 15346307
Marca: Buerkle™ 53780001
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Disposable scoop made of polystyrene is produced and assembled in a cleanroom class 7 and individually packaged for disposable use. The disposable scoop is ideal for powders, granulates, and pastes.
Scoop, disposable with or without cover
  • Polystyrene, white
  • Individually packaged
  • Production and packaging according to Cleanroom Class 7
  • With EU Foodstuffs and FDA approval
TRUSTED_SUSTAINABILITY
Specifica
Descrizione Without Cover
Capacità (sistema metrico) 25 mL
Monouso Single-use
Product Type Disposable Scoop
Include Without Cover
Materiale Polystyrene
Lunghezza (sistema metrico) 141 mm