Learn More
Smeg™ Laboratory Glassware Washer
Clean a wide range of glassware with this washer. Smeg™ Laboratory Glassware Washers are manufactured with high quality materials and feature faultless, intuitive performance. Specific accessories ensure high quality washing of even the most difficult and dirtier instruments.
Marca:  Smeg™ GW0160S
Dettagli aggiuntivi : Peso : 120.00000kg
Codice prodotto. 15354962
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Descrizione
Descrizione
  • Microprocessor electronic control
  • Active thermodynamic drying
  • Alkaline powder detergent dispenser
  • Peristaltic pump for acid liquid neutralizer
  • Washing chamber and internal door in AISI 304 stainless steel
  • External panels made in AISI 304 stainless steel
  • 2 independent washing levels through telescopic guides
  • Incorporated water softener with salt sensor
  • Cold water and demineralized water connections
  • Electronic door lock system for user safety
  • Without trolleys
Specifica
Specifica
Stainless Steel
7 default programs
490 x 525 x 570mm
Glassware Washer
Washing up to 85°C
3/N/PE 400V - 50Hz - 7 kW max