Learn More
Smeg™ Laboratory Glassware Washer
missing translation for 'orderingAttributeHoverText'
Include:
20 default programs + 10 free
20 default programs + 10 free, active thermodynamic drying
7 default programs
7 default programs, alkaline powder detergent dispenser
9 default programs + 6 free
Requisiti elettrici:
1/N/PE 230V - 50Hz - 2,8 kW max
3/N/PE 400V - 50Hz - 7 kW max
3/N/PE 400V - 50Hz - 7 kW max (switchable single-phase 230V)
Impostazione della temperatura:
Washing up to 85°C
Washing up to 95°C
missing translation for 'unitSize'
Pezzo
Description
- Microprocessor electronic control
- Active thermodynamic drying
- Alkaline powder detergent dispenser
- Peristaltic pump for acid liquid neutralizer
- Washing chamber and internal door in AISI 304 stainless steel
- External panels made in AISI 304 stainless steel
- 2 independent washing levels through telescopic guides
- Incorporated water softener with salt sensor
- Cold water and demineralized water connections
- Electronic door lock system for user safety
- Without trolleys
Specifications
Specifications
| Materiale | Stainless Steel |
| Product Type | Glassware Washer |
| Include | 9 default programs + 6 free |
| Impostazione della temperatura | Washing up to 95°C |
| Dimensioni interne (L x P x A) | 490 x 525 x 570mm |
| Requisiti elettrici | 1/N/PE 230V - 50Hz - 2,8 kW max |