Learn More
Cytiva Laboratory Manifold Standard End Cap
Click to view available options
:
For use with Laboratory Manifold
Descrizione
- Lab manifold is designed for flexibility and reduction in cross-contamination
- Lab manifold features interchangeable adapters for most funnels, and a coupling device to double testing capacity while still enabling sterilization in standard autoclaves
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Laboratory Manifold |
| Sterilità | Sterile |