Learn More
Nickel Electro™ Laboratory Stainless Steel Crucible Lid
Sfoglia Tutto Nickel-Electro Prodotti
Click to view available options
Diametro (sistema metrico) esterno:
30 mm
35 mm
40 mm
50 mm
Dimensione della confezione:
pezzi 10
Descrizione
Includes
DiameterMetricOuter
Requires
Autoclavable,dimMaterial,ProductTypeSpecs,Quantity,Shape
- Non-magnetic
- Stamped with the word Stainless Steel to assist in the identification of material when in use
- Flat shape with a depression for centering on crucibles
- Includes an extension on one side that serves as a handle
- Crevice free
Non magnetic, bright finish, crevice free lid, handle
Specifica
Specifica
| Materiale | Stainless Steel |
| Quantità | 10 Pk. |
| Autoclavabile | Autoclavable |
| Diametro (sistema metrico) esterno | 35 mm |
| Product Type | Crucible lid |
| Forma | Flat |