Learn More
Munktell Filtrak™ LabSorb Coated Surface Protection Papers
Click to view available options
Dimensioni (L x P):
500 mm x 100 m
Dimensione della confezione:
Pezzo
Descrizione
- Spills are quickly absorbed into the surface layer of cellulose material and prevented from penetrating to the surface of the work bench by a thin coating of polyethylene that serves as a carrier for the absorbent layer
- Papers can be treated with disinfectants for use in clinical laboratories to prevent biological contamination
- Also suitable for lining chemical storage areas, trays and experimental animal cages
- Water absorption of 150%
- Basis weight of 140 g/m²
- Provided individually
Specifica
Specifica
| Descrizione | Highly absorbent |
| Materiale | Cellulose, Polyethylene |
| N. per confezione | 1/pk |
| Tipo | Surface Protection Paper |
| Dimensioni (L x P) | 500 mm x 100 m |