Learn More
Sartorius™ Labsorb™ Polyethylene Coated Paper
Sfoglia Tutto Sartorius Prodotti
Click to view available options
Dimensioni (L x P):
460 x 570 mm
480 x 600 mm
Dimensione della confezione:
pezzi 50
Descrizione
- Surface protection paper
- With cellulose side up, the paper absorbs liquids, which are stopped by the polyethylene layer preventing from soaking through
- With polyethylene side up, useful for recovery of valuable or toxic liquids
Specifica
Specifica
| Descrizione | Labsorb |
| Materiale | Polyethylene-coated Paper |
| Quantità | 50 Pk. |
| Tipo | Surface Protection Paper |
| Dimensioni (L x P) | 460 x 570 mm |