missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactam of (S)-Carboxymethyl-(D,L)-cysteine, Mikromol™
Click to view available options
Quantità:
25 mg
Dimensione della confezione:
25mg
Descrizione
Lactam of (S)-Carboxymethyl-(D,L)-cysteine
Specifica
Specifica
| Materiale o nome chimico | Lactam of (S)-Carboxymethyl-(D,L)-cysteine |
| CAS | 14226-97-2 |
| Nomi componenti o analita | Lactam of (S)-Carboxymethyl-(D,L)-cysteine |
| Shipping Condition | Room Temperature |
| Formula molecolare | C5 H7 N O3 S |
| Certificazione/Conformità | ISO 17025 |
| Sinonimo | 3-Thiomorpholinecarboxylic acid, 5-oxo-, 5-Oxo-3-thiomorpholinecarboxylic acid |
| InChI Formula | InChI=1S/C5H7NO3S/c7-4-2-10-1-3(6-4)5(8)9/h3H,1-2H2,(H,6,7)(H,8,9) |
| SMILES | OC(=O)C1CSCC(=O)N1 |
| IUPAC Name | 5-oxothiomorpholine-3-carboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction