Learn More
Thermo Scientific™ Lambda Exonuclease (10 U/μL)
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Quantità:
1,000 units
5,000 units
Dimensione della confezione:
1000 unità
5000 unità
Descrizione
Thermo Scientific Lambda Exonuclease is a highly processive 5'≥3' exodeoxyribonuclease. It selectively digests the 5'-phosphorylated strand of double-stranded DNA. The enzyme exhibits low activity on single-stranded DNA and non-phosphorylated DNA, and has no activity at nicks and limited activity at gaps in DNA.
Highlights
- Active in Thermo Scientific PCR buffers
Applications
- Generating single-stranded PCR products for use in:
- DNA sequencing
- Analysis of DNA single-strand conformation polymorphism (SSCP)
- Rolling circle amplification
- Producing single-stranded DNA from double-stranded DNA fragments
- Cloning of PCR products
Note
Use of this enzyme in certain applications may be covered by patents and may require a license.
Specifica
Specifica
| Concentrazione | 10 U/μL |
| Enzyme | Exonuclease |
| Quantità | 1,000 units |
| Product Type | Exonuclease |